Tisk…

N-Methylbenzenesulphonamide 97%

Dodavatel: Apollo Scientific
Varování

OR911104-100G OR911104-25G
APOSOR911104-100GEA 10520 CZK
APOSOR911104-100G APOSOR911104-25G
N-Methylbenzenesulphonamide 97%
N-Methylbenzenesulphonamide
Vzorec: C₇H₉NO₂S
MW: 171,21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00025023
CAS číslo: 5183-78-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR