Tisk…

5-Nitro-2-(n-propylamino)pyridine 97%

Dodavatel: Apollo Scientific

Synonyms: 5-Nitro-N-propylpyridin-2-amine

OR911057-25G OR911057-5G
APOSOR911057-25GEA 13360 CZK
APOSOR911057-25G APOSOR911057-5G
5-Nitro-2-(n-propylamino)pyridine 97%
5-Nitro-2-(n-propylamino)pyridine
Vzorec: C₈H₁₁N₃O₂
MW: 181,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00084819
CAS číslo: 25948-11-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR