3-Methyl-4-(4-morpholinyl)aniline 96%
Dodavatel: Apollo Scientific
Synonyms:
3-Methyl-4-morpholinoaniline
OR910928-1G
OR910928-250MG
APOSOR910928-1GEA
2610
CZK
APOSOR910928-1G
APOSOR910928-250MG
3-Methyl-4-(4-morpholinyl)aniline 96%
3-Methyl-4-(4-morpholinyl)aniline
Vzorec:
C₁₁H₁₆N₂O Storage Temperature: Ambient |
MDL Number:
MFCD09755865 CAS číslo: 112900-82-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...