Tisk…

Methyl indoline-6-carboxylate 98%

Dodavatel: Apollo Scientific

OR910526-1G OR910526-5G
APOSOR910526-1GEA 2320 CZK
APOSOR910526-1G APOSOR910526-5G
Methyl indoline-6-carboxylate 98%
Methyl indoline-6-carboxylate
Vzorec: C₁₀H₁₁NO₂
Storage Temperature: Ambient
MDL Number: MFCD07371629
CAS číslo: 341988-36-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR