Tisk…

4-(Pyrrolidin-1-ylmethyl)benzonitrile hydrochloride 98%

Dodavatel: Apollo Scientific

Synonyms: 4-(Pyrrolidin-1-ylmethyl)benzonitrile

OR910409-1G OR910409-5G
APOSOR910409-1GEA 6220 CZK
APOSOR910409-1G APOSOR910409-5G
4-(Pyrrolidin-1-ylmethyl)benzonitrile hydrochloride 98%
4-(Pyrrolidin-1-ylmethyl)benzonitrile hydrochloride
Vzorec: C₁₂H₁₅ClN₂
MW: 222,72 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13857415
CAS číslo: 78064-96-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR