Ethyl-5-methyloxazole-4-carboxylate 95%
Dodavatel: Apollo Scientific
OR910383-1G
OR910383-250MG
OR910383-5G
OR910383-500MG
APOSOR910383-1GEA
2610
CZK
APOSOR910383-1G
APOSOR910383-250MG
APOSOR910383-5G
APOSOR910383-500MG
Ethyl-5-methyloxazole-4-carboxylate 95%
Ethyl-5-methyloxazole-4-carboxylate
Vzorec:
C₇H₉NO₃ Storage Temperature: Ambient |
MDL Number:
MFCD00105462 CAS číslo: 32968-44-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...