Tisk…

1-(4-Chlorophenyl)imidazole 96%

Dodavatel: Apollo Scientific

OR910304-1G OR910304-5G
APOSOR910304-1GEA 2150 CZK
APOSOR910304-1G APOSOR910304-5G
1-(4-Chlorophenyl)imidazole 96%
1-(4-Chlorophenyl)imidazole
Vzorec: C₉H₇ClN₂
Storage Temperature: Ambient
MDL Number: MFCD00041208
CAS číslo: 51581-54-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR