Tisk…

1-Ethyl-4-iodo-1H-pyrazole

Dodavatel: Apollo Scientific
Varování

OR910150-1G OR910150-5G
APOSOR910150-1GEA 622 CZK
APOSOR910150-1G APOSOR910150-5G
1-Ethyl-4-iodo-1H-pyrazole
1-Ethyl-4-iodo-1H-pyrazole
Vzorec: C₅H₇IN₂
Storage Temperature: Ambient
MDL Number: MFCD13195706
CAS číslo: 172282-34-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR