Tisk…

2-Methyl-2H-indazole 98%

Dodavatel: Apollo Scientific

OR910128-1G OR910128-5G
APOSOR910128-1GEA 6090 CZK
APOSOR910128-1G APOSOR910128-5G
2-Methyl-2H-indazole 98%
2-Methyl-2H-indazole
Vzorec: C₈H₈N₂
MW: 132,162 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00234418
CAS číslo: 4838-00-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR