Tisk…

3-Methyl-1-phenyl-1H-pyrazole 95%

Dodavatel: Apollo Scientific

OR910108-5G
APOSOR910108-5GEA 2320 CZK
APOSOR910108-5G
3-Methyl-1-phenyl-1H-pyrazole 95%
3-Methyl-1-phenyl-1H-pyrazole
Vzorec: C₁₀H₁₀N₂
MW: 158,2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003123
CAS číslo: 1128-54-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR