6-Methylimidazo[1,2-a]pyridine 98%
Dodavatel: Apollo Scientific
OR910055-1G
OR910055-5G
APOSOR910055-1GEA
2900
CZK
APOSOR910055-1G
APOSOR910055-5G
6-Methylimidazo[1,2-a]pyridine 98%
6-Methylimidazo[1,2-a]pyridine
Vzorec:
C₈H₈N₂ Storage Temperature: Ambient |
MDL Number:
MFCD05663806 CAS číslo: 874-38-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...