Tisk…

2,4-Dichloro-5-cyanothiazole 97%

Dodavatel: Apollo Scientific

OR910022-1G OR910022-5G
APOSOR910022-1GEA 5590 CZK
APOSOR910022-1G APOSOR910022-5G
2,4-Dichloro-5-cyanothiazole 97%
2,4-Dichloro-5-cyanothiazole
Vzorec: C₄Cl₂N₂S
Storage Temperature: Ambient
MDL Number: MFCD08669569
CAS číslo: 82554-18-5

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR