Tisk…

5,7-Dichloroquinoline 98%

Dodavatel: Apollo Scientific

OR909992-10G OR909992-1G OR909992-5G
APOSOR909992-10GEA 18420 CZK
APOSOR909992-10G APOSOR909992-1G APOSOR909992-5G
5,7-Dichloroquinoline 98%
5,7-Dichloroquinoline
Vzorec: C₉H₅Cl₂N
MW: 198,0500 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11504933
CAS číslo: 4964-77-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR