Tisk…

6,8-Dichloroquinoline 98%

Dodavatel: Apollo Scientific

OR909969-1G OR909969-5G
APOSOR909969-1GEA 5390 CZK
APOSOR909969-1G APOSOR909969-5G
6,8-Dichloroquinoline 98%
6,8-Dichloroquinoline
Vzorec: C₉H₅Cl₂N
Storage Temperature: Ambient
MDL Number: MFCD00040711
CAS číslo: 703-66-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR