Tisk…

1-Methyl-3-isoquinolinol 98%

Dodavatel: Apollo Scientific

Synonyms: 1-Methylisoquinolin-3-ol

OR909962-1G OR909962-5G
APOSOR909962-1GEA 4900 CZK
APOSOR909962-1G APOSOR909962-5G
1-Methyl-3-isoquinolinol 98%
1-Methyl-3-isoquinolinol
Vzorec: C₁₀H₉NO
Storage Temperature: Ambient
MDL Number: MFCD11053887
CAS číslo: 16535-89-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR