Tisk…

6-Chloroimidazo[1,2-a]pyridine-3-carbaldehyde 97%

Dodavatel: Apollo Scientific

OR909921-1G OR909921-5G
APOSOR909921-1GEA 3190 CZK
APOSOR909921-1G APOSOR909921-5G
6-Chloroimidazo[1,2-a]pyridine-3-carbaldehyde 97%
6-Chloroimidazo[1,2-a]pyridine-3-carbaldehyde
Vzorec: C₈H₅ClN₂O
MW: 180,5900 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09909649
CAS číslo: 29096-59-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR