5-Ethyl-3-(4-methylphenyl)-1,2,4-oxadiazole 98%
Dodavatel: Apollo Scientific
Synonyms:
5-Ethyl-3-p-tolyl-1,2,4-oxadiazole
OR909868-1G
OR909868-5G
APOSOR909868-1GEA
4140
CZK
APOSOR909868-1G
APOSOR909868-5G
5-Ethyl-3-(4-methylphenyl)-1,2,4-oxadiazole 98%
5-Ethyl-3-(4-methylphenyl)-1,2,4-oxadiazole
Vzorec:
C₁₁H₁₂N₂O Storage Temperature: Ambient |
MDL Number:
MFCD09972167 CAS číslo: 81386-31-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...