Tisk…

4-Bromo-1-propyl-1H-pyrazole 96%

Dodavatel: Apollo Scientific

OR909813-5G
APOSOR909813-5GEA 622 CZK
APOSOR909813-5G
4-Bromo-1-propyl-1H-pyrazole 96%
4-Bromo-1-propyl-1H-pyrazole
Vzorec: C₆H₉BrN₂
Storage Temperature: Ambient
MDL Number: MFCD09972141
CAS číslo: 141302-33-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR