Tisk…
4-Methoxy-N-methylaniline 98%
Dodavatel: Apollo Scientific
Synonyms: N-Methyl-p-anisidine
OR909746-25G
APOSOR909746-25GEA 1820 CZK
APOSOR909746-25G
4-Methoxy-N-methylaniline 98%
4-Methoxy-N-methylaniline
Vzorec: C₈H₁₁NO
MW: 137,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00008399
CAS číslo: 5961-59-1