Tisk…

3-Bromo-5-methyl-1H-pyrazole 98%

Dodavatel: Apollo Scientific

OR909643-1G OR909643-5G
APOSOR909643-1GEA 1950 CZK
APOSOR909643-1G APOSOR909643-5G
3-Bromo-5-methyl-1H-pyrazole 98%
3-Bromo-5-methyl-1H-pyrazole
Vzorec: C4H5BrN2
MW: 161,0000 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11215487
CAS číslo: 57097-81-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR