Tisk…

3-Amino-3-(2-pyridinyl)acrylonitrile 98%

Dodavatel: Apollo Scientific

OR909633-1G OR909633-5G
APOSOR909633-1GEA 6220 CZK
APOSOR909633-1G APOSOR909633-5G
3-Amino-3-(2-pyridinyl)acrylonitrile 98%
3-Amino-3-(2-pyridinyl)acrylonitrile
Vzorec: C₈H₇N₃
MW: 145,17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01871647
CAS číslo: 55330-52-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR