5-Acetamidoisoquinoline 98%
Dodavatel: Apollo Scientific
OR909581-1G
OR909581-5G
APOSOR909581-1GEA
6220
CZK
APOSOR909581-1G
APOSOR909581-5G
5-Acetamidoisoquinoline 98%
5-Acetamidoisoquinoline
Vzorec:
C₁₁H₁₀N₂O Storage Temperature: Ambient |
MDL Number:
MFCD05981994 CAS číslo: 27461-33-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...