Tisk…

4-tert-Butoxyaniline 96%

Dodavatel: Apollo Scientific

OR909518-1G OR909518-5G
APOSOR909518-1GEA 4300 CZK
APOSOR909518-1G APOSOR909518-5G
4-tert-Butoxyaniline 96%
4-tert-Butoxyaniline
Vzorec: C₁₀H₁₅NO
Storage Temperature: Ambient
MDL Number: MFCD11196184
CAS číslo: 57120-36-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR