1-Isopropyl-1,3-dihydro-2H-benzimidazol-2-one 98%
Dodavatel: Apollo Scientific
Synonyms:
1-Isopropyl-3H-1,3-benzodiazol-2-one
OR909495-1G
APOSOR909495-1GEA
6880
CZK
APOSOR909495-1G
1-Isopropyl-1,3-dihydro-2H-benzimidazol-2-one 98%
1-Isopropyl-1,3-dihydro-2H-benzimidazol-2-one
Vzorec:
C₁₀H₁₂N₂O MW: 176,22 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01067639 CAS číslo: 35681-40-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...