Tisk…

1,3-Dimethyl-1,3-dihydro-2H-benzimidazol-2-one 95%

Dodavatel: Apollo Scientific

Synonyms: 1,3-Dimethyl-1H-benzo[d]imidazol-2(3H)-one, 2-benzimidazolinone

OR909408-1G OR909408-5G
APOSOR909408-1GEA 1540 CZK
APOSOR909408-1G APOSOR909408-5G
1,3-Dimethyl-1,3-dihydro-2H-benzimidazol-2-one 95%
1,3-Dimethyl-1,3-dihydro-2H-benzimidazol-2-one
Vzorec: C₉H₁₀N₂O
MW: 162,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00224248
CAS číslo: 3097-21-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR