Tisk…

7-Methoxybenzofuran 95%

Dodavatel: Apollo Scientific

OR909389-1G OR909389-5G
APOSOR909389-1GEA 3770 CZK
APOSOR909389-1G APOSOR909389-5G
7-Methoxybenzofuran 95%
7-Methoxybenzofuran
Vzorec: C₉H₈O₂
MW: 148,16 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01310825
CAS číslo: 7168-85-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR