Tisk…

6-Chloro-7-methylquinoxaline 98%

Dodavatel: Apollo Scientific

OR909336-1G OR909336-25G OR909336-5G
APOSOR909336-1GEA 4140 CZK
APOSOR909336-1G APOSOR909336-25G APOSOR909336-5G
6-Chloro-7-methylquinoxaline 98%
6-Chloro-7-methylquinoxaline
Vzorec: C₉H₇ClN₂
Storage Temperature: Ambient
MDL Number: MFCD15143565
CAS číslo: 646504-79-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR