Tisk…

5-Methyl-1,3-dihydro-2H-benzimidazol-2-one 96%

Dodavatel: Apollo Scientific

Synonyms: 5-Methylbenzoimidazol-2(3H)-one

OR909301-1G OR909301-5G
APOSOR909301-1GEA 1410 CZK
APOSOR909301-1G APOSOR909301-5G
5-Methyl-1,3-dihydro-2H-benzimidazol-2-one 96%
5-Methyl-1,3-dihydro-2H-benzimidazol-2-one
Vzorec: C₈H₈N₂O
MW: 148,17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00754728
CAS číslo: 5400-75-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR