Tisk…

4-Bromo-3-isopropyl-1H-pyrazole 98%

Dodavatel: Apollo Scientific

OR909286-5G
APOSOR909286-5GEA 6290 CZK
APOSOR909286-5G
4-Bromo-3-isopropyl-1H-pyrazole 98%
4-Bromo-3-isopropyl-1H-pyrazole
Vzorec: C₆H₉BrN₂
Storage Temperature: Ambient
MDL Number: MFCD16249535
CAS číslo: 60061-60-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR