Tisk…

1-(4-Bromophenyl)cyclobutanecarboxylic acid 97%

Dodavatel: Apollo Scientific
Varování

OR909243-1G OR909243-5G
APOSOR909243-1GEA 1660 CZK
APOSOR909243-1G APOSOR909243-5G
1-(4-Bromophenyl)cyclobutanecarboxylic acid 97%
1-(4-Bromophenyl)cyclobutanecarboxylic acid
Vzorec: C₁₁H₁₁BrO₂
MW: 255,1078 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08443165
CAS číslo: 151157-49-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR