Tisk…

1-Phenylcyclohexanecarboxylic acid 97%

Dodavatel: Apollo Scientific

OR909237-1G OR909237-5G
APOSOR909237-1GEA 1740 CZK
APOSOR909237-1G APOSOR909237-5G
1-Phenylcyclohexanecarboxylic acid 97%
1-Phenylcyclohexanecarboxylic acid
Vzorec: C₁₃H₁₆O₂
Storage Temperature: Ambient
MDL Number: MFCD00037152
CAS číslo: 1135-67-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR