4,5-Dichloro-o-toluidine 98%
Dodavatel: Apollo Scientific
Synonyms:
4,5-Dichloro-2-methylaniline
OR909235-1G
OR909235-5G
APOSOR909235-1GEA
2110
CZK
APOSOR909235-1G
APOSOR909235-5G
4,5-Dichloro-o-toluidine 98%
4,5-Dichloro-o-toluidine
Vzorec:
C₇H₇Cl₂N MW: 176,0432 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11855825 CAS číslo: 2387-08-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...