2-Methoxy-N-methyaniline 95%
Dodavatel: Apollo Scientific
Synonyms:
2-Methoxy-N-methylaniline
OR909224-25G
OR909224-5G
APOSOR909224-25GEA
16140
CZK
APOSOR909224-25G
APOSOR909224-5G
2-Methoxy-N-methyaniline 95%
2-Methoxy-N-methyaniline
Vzorec:
C₈H₁₁NO MW: 137,1790 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03093822 CAS číslo: 10541-78-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...