Tisk…

3-Chloro-N-4-dimethylaniline hydrochloride 98%

Dodavatel: Apollo Scientific

OR909214-1G OR909214-5G
APOSOR909214-1GEA 4770 CZK
APOSOR909214-1G APOSOR909214-5G
3-Chloro-N-4-dimethylaniline hydrochloride 98%
3-Chloro-N-4-dimethylaniline hydrochloride
Vzorec: C₈H₁₁Cl₂N
Storage Temperature: Ambient
MDL Number: MFCD12756444
CAS číslo: 7745-94-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR