Tisk…

3-tert-Butyl-1H-pyrazole 97%

Dodavatel: Apollo Scientific

Synonyms: 3-(tert-Butyl)pyrazole

OR909140-5G
APOSOR909140-5GEA 5360 CZK
APOSOR909140-5G
3-tert-Butyl-1H-pyrazole 97%
3-tert-Butyl-1H-pyrazole
Vzorec: C₇H₁₂N₂
MW: 124,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00127337
CAS číslo: 15802-80-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR