Tisk…

7-Methyl-3-nitroimidazo[1,2-a]pyridine 98%

Dodavatel: Apollo Scientific

OR908961-10G OR908961-1G OR908961-5G
APOSOR908961-10GEA 20270 CZK
APOSOR908961-10G APOSOR908961-1G APOSOR908961-5G
7-Methyl-3-nitroimidazo[1,2-a]pyridine 98%
7-Methyl-3-nitroimidazo[1,2-a]pyridine
Vzorec: C₈H₇N₃O₂
Storage Temperature: Ambient
MDL Number: MFCD00179132
CAS číslo: 34165-07-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR