2,6-Dimethyltetrahydro-4H-pyran-4-one
Dodavatel: Apollo Scientific
|
Varování
|
OR9085-1G
OR9085-5G
APOSOR9085-1GEA
11180
CZK
APOSOR9085-1G
APOSOR9085-5G
2,6-Dimethyltetrahydro-4H-pyran-4-one
2,6-Dimethyltetrahydro-4H-pyran-4-one
Vzorec:
C₇H₁₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD01693965 CAS číslo: 1073-79-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...