2-Cyano-2-methylpropanoic acid 98%
Dodavatel: Apollo Scientific
Synonyms:
2-Cyano-2,2-dimethylacetic acid
OR908203-1G
OR908203-5G
APOSOR908203-1GEA
1820
CZK
APOSOR908203-1G
APOSOR908203-5G
2-Cyano-2-methylpropanoic acid 98%
2-Cyano-2-methylpropanoic acid
Vzorec:
C₅H₇NO₂ MW: 113,116 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11100210 CAS číslo: 22426-30-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...