Tisk…

Phenylmalonic acid 95%

Dodavatel: Apollo Scientific

OR908114-100G OR908114-25G
APOSOR908114-100GEA 1870 CZK
APOSOR908114-100G APOSOR908114-25G
Phenylmalonic acid 95%
Phenylmalonic acid
Vzorec: C₉H₈O₄
Storage Temperature: Ambient
MDL Number: MFCD00004253
CAS číslo: 2613-89-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR