Methyl-2-(chloromethyl)benzoate 95%
Dodavatel: Apollo Scientific
Synonyms:
Methyl 2-(chloromethyl)benzoate
OR908066-25G
OR908066-5G
APOSOR908066-25GEA
10150
CZK
APOSOR908066-25G
APOSOR908066-5G
Methyl-2-(chloromethyl)benzoate 95%
Methyl-2-(chloromethyl)benzoate
Vzorec:
C₉H₉ClO₂ MW: 184.6196 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06200781 CAS číslo: 34040-62-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...