4,5-Dibromothiophene-2-carboxaldehyde
Dodavatel: Apollo Scientific
|
Varování
|
OR908049-1G
OR908049-5G
APOSOR908049-1GEA
1240
CZK
APOSOR908049-1G
APOSOR908049-5G
4,5-Dibromothiophene-2-carboxaldehyde
4,5-Dibromothiophene-2-carboxaldehyde
Vzorec:
C₅H₂Br₂OS Storage Temperature: Ambient |
MDL Number:
MFCD00225242 CAS číslo: 38071-22-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...