2-(Isopropylthio)benzoic acid 98%
Dodavatel: Apollo Scientific
OR908021-1G
OR908021-25G
OR908021-5G
APOSOR908021-1GEA
3740
CZK
APOSOR908021-1G
APOSOR908021-25G
APOSOR908021-5G
2-(Isopropylthio)benzoic acid 98%
2-(Isopropylthio)benzoic acid
Vzorec:
C₁₀H₁₂O₂S MW: 196,27 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07364761 CAS číslo: 41394-95-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...