Methyl-3-ethoxybenzoate 98%
Dodavatel: Apollo Scientific
Synonyms:
Methyl 3-ethoxybenzoate
OR907890-25G
APOSOR907890-25GEA
3060
CZK
APOSOR907890-25G
Methyl-3-ethoxybenzoate 98%
Methyl-3-ethoxybenzoate
Vzorec:
C₁₀H₁₂O₃ MW: 180,2000 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00017263 CAS číslo: 108593-47-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...