N,N-Dimethyl-3-methylbenzamide 98%
Dodavatel: Apollo Scientific
Synonyms:
N,N,3-Trimethylbenzamide
OR907885-1G
OR907885-25G
OR907885-5G
APOSOR907885-1GEA
3540
CZK
APOSOR907885-1G
APOSOR907885-25G
APOSOR907885-5G
N,N-Dimethyl-3-methylbenzamide 98%
N,N-Dimethyl-3-methylbenzamide
Vzorec:
C₁₀H₁₃NO Storage Temperature: Ambient |
CAS číslo:
6935-65-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...