Tisk…

3-Methyl-N-tert-butylbenzamide 98%

Dodavatel: Apollo Scientific

Synonyms: N-tert-Butyl-3-methylbenzamide

OR907819-100G OR907819-25G OR907819-5G
APOSOR907819-100GEA 17990 CZK
APOSOR907819-100G APOSOR907819-25G APOSOR907819-5G
3-Methyl-N-tert-butylbenzamide 98%
3-Methyl-N-tert-butylbenzamide
Vzorec: C₁₂H₁₇NO
Storage Temperature: Ambient
MDL Number: MFCD00996285
CAS číslo: 42498-33-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR