(±)-α-Chlorophenylacetic acid 98%
Dodavatel: Apollo Scientific
Synonyms:
2-Chloro-2-phenylacetic acid
OR907811-10G
OR907811-1G
OR907811-5G
APOSOR907811-10GEA
8270
CZK
APOSOR907811-10G
APOSOR907811-1G
APOSOR907811-5G
(±)-α-Chlorophenylacetic acid 98%
(±)-α-Chlorophenylacetic acid
Vzorec:
C₈H₇ClO₂ MW: 170.5900 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00037753 CAS číslo: 4755-72-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...