Tisk…

(4-Methylpiperidin-4-yl)methanol 95%

Dodavatel: Apollo Scientific

OR907789-1G OR907789-250MG
APOSOR907789-1GEA 4270 CZK
APOSOR907789-1G APOSOR907789-250MG
(4-Methylpiperidin-4-yl)methanol 95%
(4-Methylpiperidin-4-yl)methanol
Vzorec: C₇H₁₅NO
MW: 129,2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09991910
CAS číslo: 297172-16-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR