6-Bromo-1,2,3,4-tetrahydroisoquinoline hydrochloride 95%
Dodavatel: Apollo Scientific
|
Varování
|
OR907336-10G
OR907336-5G
APOSOR907336-10GEA
0
CZK
APOSOR907336-10G
APOSOR907336-5G
6-Bromo-1,2,3,4-tetrahydroisoquinoline hydrochloride 95%
6-Bromo-1,2,3,4-tetrahydroisoquinoline hydrochloride
Vzorec:
C₉H₁₁BrClN MW: 248,5473 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08234662 CAS číslo: 215798-19-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...