(4-Bromothiazol-5-yl)methanol 95%
Dodavatel: Apollo Scientific
OR907317-1G
OR907317-250MG
APOSOR907317-1GEA
2440
CZK
APOSOR907317-1G
APOSOR907317-250MG
(4-Bromothiazol-5-yl)methanol 95%
(4-Bromothiazol-5-yl)methanol
Vzorec:
C₄H₄BrNOS MW: 194,0497 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09909724 CAS číslo: 262444-15-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...