Tisk…

2,5-Dichloro-3-thenoic acid 97%

Dodavatel: Apollo Scientific

Synonyms: 2,5-Dichlorothiophene-3-carboxylic acid

OR907296-100G OR907296-25G
APOSOR907296-100GEA 8670 CZK
APOSOR907296-100G APOSOR907296-25G
2,5-Dichloro-3-thenoic acid 97%
2,5-Dichloro-3-thenoic acid
Vzorec: C₅H₂Cl₂O₂S
MW: 197,04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00082908
CAS číslo: 36157-41-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR